F.A.M.E. Mix GLC-50, certified reference material, TraceCERT®, Supelco®
Leverandør: Merck
SUPL1894-1AMPEA 430 NOK
SUPL1894-1AMP
F.A.M.E. Mix GLC-50, certified reference material, TraceCERT®, Supelco®
Methyl cis-13-docosenoate 25 wt. %; Methyl cis-11-eicosenoate 25 wt. %; Methyl oleate 25 wt. %; Methyl palmitoleate 25 wt. %
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. 