F.A.M.E. Mix GLC-70, certified reference material, TraceCERT®, Supelco®
Leverandør: Merck
SUPL1897-1AMPEA 795 NOK
SUPL1897-1AMP
F.A.M.E. Mix GLC-70, certified reference material, TraceCERT®, Supelco®
This quantitative mix is useful for determining relative retention times and approximating response factors.
Methyl decanoate 20 wt. %; Methyl dodecanoate 20 wt. %; Methyl nonanoate 20 wt. %; Methyl octanoate 20 wt. %; Methyl undecanoate 20 wt. %