These single use overalls are made from lightweight, breathable anti-static SMS fabric.
- Lightweight, breathable material helps to minimise the risk of heat stress
- Silicone-free: critical in spray painting applications
- Low linting to reduce the risk of contamination in critical areas
- Optimised body fit improves wearer comfort with knitted cuffs
- Protects against sudden discharges of electrostatic energy
- Superior repellency for enhanced protection against liquids, particularly oils and alcohols compared to traditional ‘SMS’ technology
Typical applications include: Solvent degreasing and parts cleaning, handling low hazard liquids, blending filtering compounding raw materials, preparing paints, inspecting machinery and equipment for defects.
Mostly used in industrial areas such as aerospace, automotive, chemical, energy, life sciences, metal fabrication.
Sertifiseringer: Type 5/6, EN 1073-2, EN 1149-5.