Supelco® Organic Reference Standard, β-Amyrin
09236-10MG-F
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:559-70-6
- Description:β-Amyrin
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00017381
- M-Clarity quality level:MQ100
- Synonym(s):β-Amyrenol; Olean-12-en-3β-ol
- Formula:C₃₀H₅₀O
- Molecular weight:426,72
- Flash point:Not applicable