Norway
Order Entry
Thermometer adapter, Wheaton®
Thermometer adapter, Wheaton®
  WHEA165996
undefined
Thermometer adapter, Wheaton®
  WHEA165996
 : 165996
 : WHEA165996
kr 1 925,00
  1 * 1 Items
  • Description:
    Thermometer adapter, 20-400
  • Pk:
    1
This adapter has an O-ring that holds a thermometer or gas inlet tube securely. A grease-free WHEATON Connection® on one end provides an inert connection with external-threaded glass components.
The adapter can also be used with gas inlet tubing and will accommodate any tube with an OD of 5 to 7 mm.