Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Acebutolol hydrochloride, analytical standard, Supelco®
A3669-1G
:
Merck
:
Expand image
Acebutolol hydrochloride, analytical standard, Supelco®
A3669-1G
:
Merck
:
kr 405,00
1 * 1 g
-
+
Pk:
1 g
Standards:
analytical standard
CAS No.:
34381-68-5
Description:
Acebutolol hydrochloride
Description:
Acebutolol hydrochloride