Natriumtiosulfat løsning 1 mol/l (1 N), Titripur®, Supelco®


M-Clarity™ quality level = MQ100

1.60312.1003EA 1250 NOK
1.60312.1003 1.60312.1000
Natriumtiosulfat løsning 1 mol/l (1 N), Titripur®, Supelco®
Natriumtiosulfat

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formel: Na₂S₂O₃
MW: 158,11 g/mol
MDL nummer: MFCD00003499
CAS-nummer: 7772-98-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR