N-Desmethylcitalopram-D3 hydrochloride solution, certified reference material, Supelco®
Leverandør: Merck
Cerilliant®
D-074-1MLEA 2425 NOK
D-074-1ML
N-Desmethylcitalopram-D3 hydrochloride solution, certified reference material, Supelco®
Standarder Organiske standarder
A stable-labeled internal standard of N-Desmethylcitalopram, an active metabolite of the SSRI antidepressant drug citalopram.
Forpakning: Snap-N-Spike®/Snap-N-Shoot®