Absorbent granules, SpillFix
Leverandør: Brady
BRDY149060EA 23200 NOK
BRDY149060 BRDY149063 BRDY149116
Absorbent granules, SpillFix
SpillFix coconut coir granular is a completely natural producing no carcinogens or dust.
  • Premium absorbent made of 100% recycled material
  • Extremely effective for the absorption of hydrocarbons, water-based fluids, and most aggressive chemicals
  • Certified safe for landfill disposal
With more than four times the absorption power of clay granular, this product is perfect for both indoor and outdoor use.