tert-Amyl methyl ether, GPR RECTAPUR® METHOXYPENTANE™
|
Danger
|
Synonymer:
METHOXYPENTANE™
METHOXYPENTANE™ is a novel high quality ether solvent developed by INEOS Oligomers for demanding applications in the manufacture of high value fine chemicals, advanced life science intermediates, API’s, and high-tech chemicals. Methoxypentane™ is ultra-pure with a 2-methoxy-2-methyl butane content typically exceeding 99.5 wt.-%.
METHOXYPENTANE™ offers a range of unique solvent properties which helps to support user creativity in the identification of new applications for the product. The novel solvent enables researchers in academic and industrial settings to invent and design new synthesis processes and purification procedures. Furthermore, it allows process development professionals in industry to establish efficient manufacturing routes and production managers to achieve and improve selectivity and product yield while reducing waste.
NB: METHOXYPENTANE™ is a trademark of INEOS Oligomers.
Formel:
C₆H₁₄O MW: 102,17 g/mol Kokepunkt: 85-86 °C (lit.) Tetthet: 0,76-0.78 g/mL (25 °C) (lit.) Flash punkt: -7 °C |
MDL nummer:
MFCD00010186 CAS-nummer: 994-05-8 EINECS: 213-611-4 FN: 3271 ADR: 3,II |
Spesifikasjon Testresultater
Sum of C6-ethers | Min. 99,8 % |
2-Methoxy-2-methylbutane | Min. 99,5 % |
Sum of C5-hydrocarbons | Max. 100 ppm |
Sum of C7-ethers | Max. 0,05 % |
2-Methyl-2-butanol | Max. 0,05 % |
Colouration (Hazen) | Max. 5 APHA |
Benzene | Max. 70 ppm |
Water | Max. 0,05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...