Surgical mask, ear loop, type II
Leverandør: 3M Medical
MMMEM1826EA 2050 NOK
MMMEM1826
Surgical mask, ear loop, type II
Masker Munnbind, kirurgi
The 1826 Surgical Mask offers 98% bacterial filtration efficiency for maximum protection.
  • Provides maximum wearer comfort for extended use
  • Soft and conformable aluminum nose band
  • Mask measures 190 x 80mm
Surgical masks help to reduce potential exposure of the wearer to blood and body fluids.
Sertifiseringer: EN14683:2004