Norway
Order Entry
VWR®, Cold Cranking Simulator (CCS) viscosity standards
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier:  VWR Chemicals
Certificates
Related Information
Product Information
  • Catalog #
    No.
    Type
    Viscosity
    Pack type
    Test method
    Pk Info
    Availability
    Price
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL08
    (–35,00 °C) 857,1 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL09
    (–35,00 °C) 1175 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL10
    (–30,00 °C) 896,2 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL11
    (–30,00 °C) 1035 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL12
    (–25,00 °C) 770,4 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL13
    (–25,00 °C) 980,1 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL14
    (–20,00 °C) 826,1 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL15
    (–20,00 °C) 921,0 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL16
    (–20,00 °C) 1186 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL17
    (–15,00 °C) 872,6 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL19
    (–15,00 °C) 1075 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL20
    (–10,00 °C) 870,7 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL22
    (–10,00 °C) 1030 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL24
    (–10,00 °C) 1211 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL25
    (–10,00 °C) 1376 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL26
    (–10,00 °C) 1680 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL28
    (–10,00 °C) 2016 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL30
    (–10,00 °C) 2431 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL32
    (–10,00 °C) 3049 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL34
    (–10,00 °C) 3431 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL38
    (–10,00 °C) 4199 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL42
    (–10,00 °C) 4969 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL48
    (–10,00 °C) 6135 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    99399.260
    CL53
    (–10,00 °C) 7440 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL60
    (–10,00 °C) 9303 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL68
    (–10,00 °C) 11417 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
  • CCS Viscosity Standard
    CCS Viscosity Standard
    CL74
    (–10,00 °C) 12567 mPa s
    Glass bottle
    ASTM D5293/SAE Specification J300
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 1 175,00
    kr 1 175,00
    1 * 500 mL
       kr 1 175,00
Sets
Product Information
  • Catalog #
    No.
    Type
    Viscosity
    Pack type
    Test method
    Pk Info
    Availability
    Price
  • CCS Viscosity Standard Kit 8
    CCS Viscosity Standard Kit 8
    99403.001
    VIS-CCS-8
    8 Set: CL14, CL19, CL22, CL25, CL28, CL32, CL48, CL68.
    Glass bottle
    ASTM D5293/SAE Specification J300
    8 x 500 ml
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - kr 7 900,00
    kr 7 900,00
    1 * 1 Each
       kr 7 900,00
  • CCS Viscosity Standard Kit 14
    CCS Viscosity Standard Kit 14
    99404.001
    VIS-CCS-14
    14 Set: CL10, CL12, CL14, CL16, CL19, CL22, CL25 (2), CL28, CL32, CL38, CL48, CL60, CL68.
    Glass bottle
    ASTM D5293/SAE Specification J300
    14 x 500 ml
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - kr 13 375,00
    kr 13 375,00
    1 * 1 Each
       kr 13 375,00
  • CCS Viscosity Standard Kit 18
    CCS Viscosity Standard Kit 18
    99405.001
    VIS-CCS-18
    18 Set: CL08, CL09, CL10, CL11, CL12, CL13, CL14, CL15, CL16, CL19, CL22, CL25(x2), CL28, CL32, CL38, CL48 & CL60.
    Glass bottle
    ASTM D5293/SAE Specification J300
    18 x 500 ml
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - kr 13 900,00
    kr 13 900,00
    1 * 1 Each
       kr 13 900,00