Disodium metasilicate pentahydrate
ROTH7370.1
:
- Pack type:Plastic bottle for solids
- Pk:2,5 kg
:
:212.14 g/mol
:13,08716
:72.2 °C
:1.749 g/cm³ (20 °C)
:229-912-9
:3253
:8,III
:GHS05
:GHS07
:10213-79-3
:Na₂SiO₃·5H₂O
:MFCD00149176