
Alizarin cyanin green F
TCIAA0610-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Dark green to Dark blue to Black powder to crystal
- Lambda max.:639.0 to 643.0 nm (H2O)
- Absorbance (E1%1cm):min. 150 (H2O, 639.0 to 643.0 nm)
:
:622.57 g/mol
:14,00252
:224-546-6
:GHS07
:4403-90-1
:C₂₈H₂₂N₂O₈S₂
:MFCD00001193