Norway
Order Entry
2-(4-Bromobutoxy)tetrahydro-2H-pyran ≥98.0% (by GC)
  TCIAB3186-5G
 :  TCI
 :  
2-(4-Bromobutoxy)tetrahydro-2H-pyran ≥98.0% (by GC)
  TCIAB3186-5G
 :  TCI
 :  
 
kr 2 775,00
   
  1 * 5 g

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :237.14 g/mol
 :Freezer
 :31608-22-7
 :C₉H₁₇BrO₂
 :MFCD06654117