3-Cyanopyridine-N-oxide ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
|
Warning
|
TCIAC3416-1GEA
450
NOK
TCIAC3416-1G
3-Cyanopyridine-N-oxide ≥98.0% (by GC, titration analysis)
3-Cyanopyridine-N-oxide
Formel:
C₆H₄N₂O MW: 120,11 g/mol |
MDL nummer:
MFCD00129037 CAS-nummer: 14906-64-0 EINECS: 238-975-1 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 175.0 to 179.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...