Norway
Order Entry
 
 
2,3-Epoxynorbornane ≥95.0% (by GC)
  TCIAE0779-5G
 :  TCI
 :  
 
 
2,3-Epoxynorbornane ≥95.0% (by GC)
  TCIAE0779-5G
 :  TCI
 :  
 : E0779-5G
 : TCIAE0779-5G
 
kr 2 150,00
   
  1 * 5 g

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    124.0 to 128.0 deg-C

 

 

 :
 :110.16 g/mol
 :205-995-7
 :1325
 :4.1,II
 :GHS02
 :GHS07
 :278-74-0
 :C₇H₁₀O
 :MFCD00798067