3-Formyl-6-isopropylchromone ≥98.0% (by GC)
Leverandør: TCI EUROPE
|
Warning
|
TCIAF0856-1GEA
690
NOK
TCIAF0856-1G
3-Formyl-6-isopropylchromone ≥98.0% (by GC)
3-Formyl-6-isopropylchromone
Formel:
C₁₃H₁₂O₃ MW: 216,2326 g/mol |
MDL nummer:
MFCD00192183 CAS-nummer: 49619-58-1 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 97.0 to 101.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...