L-Leucyl-L-tyrosine
: TCI
About this item
- Specific rotation [a]20/D: 23° (C=2, H₂O)
- EINECS: 213-527-8
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Leu-Tyr-OH
- CAS No.:968-21-8
- Form:White to almost white powder to crystal
- MDL No.:MFCD00020195
- Protein synonyms:H-Leu-Tyr-OH
- Protein/peptide name:L-Leucyl-L-tyrosine
- Purity:>98,0% (T)
- Molecular weight:294,35 g/mol
- Molecular formula:C₁₅H₂₂N₂O₄
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- TCIAL0040-100MGL0040-100MG100 mg1 * 100 mgkr 245,00: L0040-100MG: TCIAL0040-100MG
- : L0040-1G: TCIAL0040-1G