
Thiosemicarbazide hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAT0222-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Iodometric Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:127.59 g/mol
:2811
:6.1,III
:GHS06
:4346-94-5
:CH₅N₃S
:MFCD00060206