These impact- and cut-resistant gloves deliver high cut and impact protection for hazardous and unpredictable environments.
- Advanced bumper design provides certified impact resistance
- HPPE liner helps keep hands cool and comfortable while providing cut resistance
- Enhanced oil grip reduces hand fatigue and injury by increasing control while easing the force required to safely handle items
- Double dip nitrile coating technology provides protection from oils/liquids and abrasion
- HIVIZ™ design increases awareness of users for safety
The glove with double-dip nitrile coating provides excellent grip and control when working with oils and other liquids. The orange HIVIZ™ design increases awareness for user safety.
Recommended for: Engagement and release of drill string slips, drilling assembly and completion, handling jagged metal, production services / processing, drilling / extraction, Heavy Equipment Servicing, transport & logistics.
Sertifiseringer: EN 388:2016 (4X43CP); Cat. II
Forpakning: 12 pairs per bag