Industrial gloves, HyFlex® 11-928
Leverandør: Ansell
HyFlex®
ANSE11-928/8EA 3050 NOK
ANSE11-928/8 ANSE11-928/9 ANSE11-928/7 ANSE11-928/10 ANSE11-928/11
Industrial gloves, HyFlex® 11-928
Hansker
This glass-free INTERCEPT liner provides increased cut protection (cut level 4).
  • Silicone and DMF-free
  • Grip performance for safe handling in dry to light oily applications
  • Double dipped nitrile coating protects against oil and industrial fluid exposure while providing high durability and extended use life
  • INTERCEPT aramid liner for increased cut performance
  • Double layer RIPEL™ nitrile coating keeps hands dry and comfortable when handling oily parts and tools
  • Intermittent contact heat protection when handling hot parts up to 212 °F / 100 °C (Contact Heat Protection Level 1)
Recommended for: General maintenance; handling sharp or irregular metal pieces; handling of glass sheets, sharpedged objects; grinding; machining / stamping / cutting
Sertifiseringer: EN 388:2016 (4X32D); EN407 (X1XXXX); Cat. III
Forpakning: 12 pairs per bag.