Chemical resistant overalls, MICROGARD® 2500 PLUS AIRline, Model 750
Leverandør: Ansell
Microgard® 2500 Plus
ANSEWH25T-00750-07EA 0 NOK
ANSEWH25T-00750-07 ANSEWH25T-00750-06 ANSEWH25T-00750-04 ANSEWH25T-00750-02 ANSEWH25T-00750-03 ANSEWH25T-00750-05
Chemical resistant overalls, MICROGARD® 2500 PLUS AIRline, Model 750
Engangsplagg
Encapsulated chemical protective suits for use with air-fed/supplied systems.
  • Providing protection from respiratory and skin hazards
  • Panoramic visor design ensures a good field of vision for the wearer
  • Unique material offering exceptional mechanical strength, liquid and particulate protection
  • Double elasticated cuffs ensure a liquid-tight connection with chemical protective gloves
  • Air permeable SMS collar maintains sufficient air in the breathing zone, allowing excess air to flow into suit body
  • Full range of movement without risk of excessive pressure causing harm to the suit or the wearer
  • Emergency rip cord permits rapid suit removal in cases of emergency
Anti-static, tested according to EN 1149-5. Attached socks with elasticated boot overflap.
Fully encapsulated suit designed for use in combination with continuous flow compressed air line breathing apparatus; Sundström SR507 (sold separately). Suit features include double elasticated cuffs, attached socks with elasticated boot overflap, panoramic visor design, four exhalation valves, air permeable SMS collar, and emergency rip cord. Stitched and taped seams. Garments are manufactured from non-woven microporous polypropylene laminate.
Recommended for transferring liquids and solids, unexpected leakages spills or other releases, plant disinfection and sanitization, tank cleaning, emergency response, spill or leakage cleanup, maintenance, and contaminated environments.
Sertifiseringer: EN 1073-1:1998; EN 1149-5:2008; Category III
Forpakning: Packaged 5 pieces/carton.