Duckbill face mask, FFP2
111-2487EA 25150 NOK
111-2487
Duckbill face mask, FFP2
Masker Munnbind
Disposable filtering half-mask acts as a respiratory protective device to protect against particles.
  • Cleanroom compatible
  • Comfortable design for extended wear
  • Low breathing resistance
  • Adjustable nose bridge
  • Bacterial filtration efficiency is greater than 95%
Disposable duck bill face mask features a six-layer material construction, comprised of spunbonded polypropylene inner and outer layers, microfibre lining fabric, and three layers of melt-blown polypropylene filter media. Additional components include a plastic nose-piece, sponge nasal pad, and synthetic rubber (latex-free) head strap.
Colour: White.
Sertifiseringer: EN 149:2001+A1:2009