Avantor® Hichrom HI-PAH, GC Columns
: Avantor
Application specifc column for polycyclic aromatics hydrocarbons.
Max temperature limits may change depending on the film thickness.
- Catalog No:
- HI48-18-010-20
- HI48-25-025-30
- HI48-25-015-30
- Phase name:
- HI-PAH
- HI-PAH
- HI-PAH
- Phase composition:
- Proprietary Phase developed for PAHs analysis
- Proprietary Phase developed for PAHs analysis
- Proprietary Phase developed for PAHs analysis
- Crossbond:
- Yes
- Yes
- Yes
- Temperature limit:
- 350 °C
- 350 °C
- 350 °C
- Column FormatFilm thicknessØ int.LengthPk
- GC ColumnGC ColumnHI48-18-010-20Capillary0,10 µm0,18 mm20 m11 * 1 Itemskr 13 225,00: HIGCHI48-18-010-20
Accessories
Specifications
Environmentally PreferablePhase nameHI-PAHPhase compositionProprietary Phase developed for PAHs analysisCrossbondYesTemperature limit350 °C - GC ColumnGC ColumnHI48-25-025-30Capillary0,25 µm0,25 mm30 m11 * 1 Itemskr 13 225,00: HIGCHI48-25-025-30
Accessories
Specifications
Environmentally PreferablePhase nameHI-PAHPhase compositionProprietary Phase developed for PAHs analysisCrossbondYesTemperature limit350 °C - GC ColumnGC ColumnHI48-25-015-30Capillary0,15 µm0,25 mm30 m11 * 1 Itemskr 13 225,00: HIGCHI48-25-015-30
Accessories
Specifications
Environmentally PreferablePhase nameHI-PAHPhase compositionProprietary Phase developed for PAHs analysisCrossbondYesTemperature limit350 °C