Norway
Order Entry
Iridium AAS Standards, (CRMs), Fluka™
Iridium AAS Standards, (CRMs), Fluka™
Certificates
Iridium AAS Standards, (CRMs), Fluka™
Product Information
  • Catalog #
    Supplier #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • HONCE4036-100ML
    E4036-100ML
    Iridium
    Ir
    1000 mg/L
    10% HCl
    Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - kr 2 500,00
    kr 2 500,00
    1 * 100 mL
       kr 2 500,00
  • HONCE4036-500ML
    E4036-500ML
    Iridium
    Ir
    1000 mg/L
    10% HCl
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - kr 8 525,00
    kr 8 525,00
    1 * 500 mL
       kr 8 525,00