Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
Order Entry
Home
Head Protection
Safety Helmets
Safety helmet, Endurance, PS55
Safety helmet, Endurance, PS55
Safety helmet, Endurance, PS55
PRTWPS55WHR
:
Portwest
Safety helmet, Endurance, PS55
PRTWPS55WHR
:
Portwest
kr 310,00
1 * 1 Items
-
+
Colour:
White
Description:
Safety helmet, Endurance
Model:
PS55
Pk:
1
Chin strap
Description
Model
Colour
Pk
PRTWPA48BKR
Chin strap for PW54/PS54/PW55/PS55/PC55 helmets
PA48
Black
5
1 * 5 Items
1 * 5 Items - kr 1 200,00
kr 1 200,00
-
+
-
+