Spinner flasks with angled sidearms, Corning®
: Corning
Borosilicate glass with side arms, non sterile.
- Baffles enhance aeration and agitation of flask contents
- Impeller design ensures optimal stirring
- Complete with impeller and caps
- Side arm design permit easy access of 25 and 50 ml pipettes
- CapacityNeckNo. of side armsPk
- 4500-125125 mlCentre neck 70 mm, side arm neck 32 mm211 * 1 Itemskr 8 875,00
Specifications
Environmentally Preferable - 4500-15L15000 mlCentre neck 100 mm, side arm neck 45 mm211 * 1 Itemskr 27 900,00
Specifications
Environmentally Preferable - 4500-1L1000 mlCentre neck 100 mm, side arm neck 45 mm211 * 1 Itemskr 14 000,00
Specifications
Environmentally Preferable - 4500-250250 mlCentre neck 70 mm, side arm neck 32 mm211 * 1 Itemskr 9 225,00
Specifications
Environmentally Preferable - 734-19114500-36L36000 mlCentre neck 100 mm, side arm neck 45 mm211 * 1 Itemskr 40 925,00
Specifications
Environmentally Preferable - 4500-3L3000 mlCentre neck 100 mm, side arm neck 45 mm211 * 1 Itemskr 15 850,00
Specifications
Environmentally Preferable - 4500-500500 mlCentre neck 100 mm, side arm neck 45 mm211 * 1 Itemskr 9 750,00
Specifications
Environmentally Preferable - 4500-6L6000 mlCentre neck 100 mm, side arm neck 45 mm211 * 1 Itemskr 18 675,00
Specifications
Environmentally Preferable - 4500-8L8000 mlCentre neck 100 mm, side arm neck 45 mm211 * 1 Itemskr 21 600,00
Specifications
Environmentally Preferable - 734-19164502-15L15000 mlCentre neck 120 mm, side arm neck 45 mm211 * 1 Itemskr 28 150,00
Specifications
Environmentally Preferable - 4502-36L36000 mlCentre neck 120 mm, side arm neck 45 mm211 * 1 Itemskr 42 175,00
Specifications
Environmentally Preferable - 4502-3L3000 mlCentre neck 120 mm, side arm neck 45 mm211 * 1 Itemskr 16 525,00
Specifications
Environmentally Preferable - 734-19194502-6L6000 mlCentre neck 120 mm, side arm neck 45 mm211 * 1 Itemskr 19 925,00
Specifications
Environmentally Preferable - 4502-8L8000 mlCentre neck 120 mm, side arm neck 45 mm211 * 1 Itemskr 22 475,00
Specifications
Environmentally Preferable