Human Umbilical vein endothelial cells (HUVEC), Corning®
Leverandør: Corning
734-1120EA 9275 NOK
BDAA354151
Human Umbilical vein endothelial cells (HUVEC), Corning®
HUVEC-2 cells are qualified for use in Corning® BioCoat™ angiogenesis systems: endothelial cell migration
The cells may be used in Corning BioCoat endothelial cell invasion and tube formation assays. They are used as model for studying endothelial differentiation and modulation of endothelial tube formation by antiangiogenic agents.
Bestillingsinformasjon: Includes Corning®Human Umbilical Vein Endothelial Cells, HUVEC-2 Cells (>5×105 Cells).