Blotting membranes, Amersham™ Hybond™-NX
Leverandør: Cytiva
RPN203TEA 3350 NOK
GEHERPN203T
Blotting membranes, Amersham™ Hybond™-NX
Amersham™ Hybond™-NX is a neutral nylon membrane with all the properties of Hybond™-N. It can be used for multiple re-probing due to its high tensile strength. Hybond™-NX gives cleaner backgrounds than Hybond™-N when using hybridisation buffer volumes below 70 µl/cm² of membrane.
  • Developed specifically for use with low hybridisation buffer volumes and high-throughput applications
Pore size: 0,45 µm
Nucleic acid size required: >50 bp