Norway
Order Entry
Latex gloves, Vigor®
undefined
Latex gloves, Vigor®
  • Catalog No:
  • 112-6139
  • 112-6138
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex
  • Latex
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Rolled
  • Rolled
  • Finger thickness:
  • 0,08 mm
  • 0,08 mm
  • Palm thickness:
  • 0,08 mm
  • 0,08 mm
  • Tensile strength:
  • 14 MPa
  • 14 MPa
  • Ultimate elongation:
  • 500%
  • 500%
  • EN norm:
  • EN 455, EN ISO 374-1, EN 374-2, EN 374-4, EN 374-5, EN 16523-1
  • EN 455, EN ISO 374-1, EN 374-2, EN 374-4, EN 374-5, EN 16523-1
  • Food contact approved:
  • +
  • +
  •  
    Model
    Glove size
    Colour
    Length
    Pk
  • 112-6138
    7026387
    LFSD Latex Vigor Bird
    S
    White
    240 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 295,00
    kr 295,00
    1 * 100 Items
       kr 295,00
    Specifications
    Ambidextrous / hand-specific
    Ambidextrous
    Environmentally Preferable
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Finish
    Textured fingertips
    Cuff style
    Rolled
    Finger thickness
    0,08 mm
    Palm thickness
    0,08 mm
    Tensile strength
    14 MPa
    Ultimate elongation
    500%
    EN norm
    EN 455, EN ISO 374-1, EN 374-2, EN 374-4, EN 374-5, EN 16523-1
    Food contact approved
    +
  • 112-6139
    7026388
    LFSD Latex Vigor Bird
    M
    White
    240 mm
    100
    1 * 100 Items
    1 * 100 Items - kr 295,00
    kr 295,00
    1 * 100 Items
       kr 295,00
    Specifications
    Ambidextrous / hand-specific
    Ambidextrous
    Environmentally Preferable
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Finish
    Textured fingertips
    Cuff style
    Rolled
    Finger thickness
    0,08 mm
    Palm thickness
    0,08 mm
    Tensile strength
    14 MPa
    Ultimate elongation
    500%
    EN norm
    EN 455, EN ISO 374-1, EN 374-2, EN 374-4, EN 374-5, EN 16523-1
    Food contact approved
    +