HPLC waste-sets, V2.0
: S.C.A.T.
Sets
- VolumeTypeMaterialPk
- HPLC waste-set 1, V2.0, GL45HPLC waste-set 1, V2.0, GL45SCTV3073073073075 LSafety canisterHDPE1 SET1 * 1 Eachkr 3 025,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 2, V2.0, GL45HPLC waste-set 2, V2.0, GL45SCTV30731030731010 LSafety canisterHDPE1 SET1 * 1 Eachkr 3 025,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 9, V2.0, S60/61HPLC waste-set 9, V2.0, S60/61SCTV30737030737020 LSafety canisterHDPE11 * 1 Itemskr 3 300,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 11, V2.0, S60/61, UN-XHPLC waste-set 11, V2.0, S60/61, UN-XSCTV3073713073715 LSafety canisterHDPE11 * 1 Itemskr 2 975,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 8, V2.0, S51, UN-XHPLC waste-set 8, V2.0, S51, UN-XSCTV3073693073695 LSafety canisterHDPE11 * 1 Itemskr 2 975,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 10, V2.0, S90HPLC waste-set 10, V2.0, S90SCTV30732430732410 LSafety canisterHDPE11 * 1 Itemskr 3 500,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 3, V2.0, S50HPLC waste-set 3, V2.0, S50SCTV3073283073285 LFlat canisterPP11 * 1 Itemskr 2 900,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 4, V2.0, S60/61HPLC waste-set 4, V2.0, S60/61SCTV30735530735512 LSafety canisterHDPE11 * 1 Itemskr 2 975,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 12, V2.0, S70/71HPLC waste-set 12, V2.0, S70/71SCTV30737230737220 LSafety canisterHDPE11 * 1 Itemskr 3 075,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 5; V2.0, GL45HPLC waste-set 5; V2.0, GL45SCTV3073663073662,5 LSafety canisterHDPE11 * 1 Itemskr 2 450,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 6, V2.0, S55HPLC waste-set 6, V2.0, S55SCTV3073673073675 LSafety canisterHDPE11 * 1 Itemskr 2 975,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 7, V2.0, S55, UN-XHPLC waste-set 7, V2.0, S55, UN-XSCTV3073683073685 LSafety canisterHDPE11 * 1 Itemskr 2 975,00
Accessories
Specifications
Environmentally Preferable
Sets, electrostatic conductive
- VolumeTypeMaterialPk
- HPLC waste-set 17, V2.0, S60/61HPLC waste-set 17, V2.0, S60/61SCTV30737730737720 LSafety canisterHDPE/PTFE11 * 1 Itemskr 4 200,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 16, V2.0, S60/61HPLC waste-set 16, V2.0, S60/61SCTV30737630737610 LSafety canisterHDPE/PTFE11 * 1 Itemskr 3 850,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 13, V2.0, S50HPLC waste-set 13, V2.0, S50SCTV3073733073735 LSpace saving canisterHDPE/PTFE11 * 1 Itemskr 5 600,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 14, V2.0, S50HPLC waste-set 14, V2.0, S50SCTV30737430737410 LSafety canisterHDPE/PTFE11 * 1 Itemskr 4 375,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 15, V2.0, S60/61HPLC waste-set 15, V2.0, S60/61SCTV3073753073755 LSafety canisterHDPE/PTFE11 * 1 Itemskr 3 775,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 18, V2.0, S60/61HPLC waste-set 18, V2.0, S60/61SCTV30737830737830 LSafety canisterHDPE/PTFE11 * 1 Itemskr 4 725,00
Accessories
Specifications
Environmentally Preferable - HPLC waste-set 19, V2.0, S90HPLC waste-set 19, V2.0, S90SCTV30737930737910 LSafety canisterHDPE/PTFE11 * 1 Itemskr 5 700,00
Accessories
Specifications
Environmentally Preferable
Safety waste caps
- SeqDescriptionThreadConnectorPk
- 34SafetyWasteCap, for space saving canisterS502× 2,3/3,2 mm Ext.Ø, 1× 5,0 - 11,5 mm Int.Ø1
Canister
- SeqDescriptionCapacityColourThreadTypeW×D×HPk
- 02Safety canister5 LNaturalGL 45UN-Y approval150×195×250 mm1
Exhaust filter
- SeqDescriptionPk
- 04Exhaust filter M, V2.0, with splash guard and change indicator, service life 6 months1