Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
/
Norway
Products
Services
Digital Solutions
Applications
Support
About
kr 0,00
Order Entry
Home
Wipes
Disposable Wipes and Towels
Paper Towels
Hand towel roll, blue, SCOTT® ESSENTIAL™
Hand towel roll, blue, SCOTT® ESSENTIAL™
Hand towel roll, blue, SCOTT® ESSENTIAL™
KIMB6692
:
KIMBERLY CLARK
Hand towel roll, blue, SCOTT® ESSENTIAL™
KIMB6692
:
KIMBERLY CLARK
: 6692
: KIMB6692
kr 4 325,00
1 * 6 Rull
-
+
Colour:
Blue
Description:
Hand towel roll
L×W:
350000×198 mm
Pk:
6 Roll