Moisture analyzer, Mark3
Leverandør: Sartorius
SARBLMA100-SET-PEA 0 NOK
SARBLMA100-SET-P
Moisture analyzer, Mark3
Analysatorer Fuktanalyse
Mark3 Moisture Analyzers for every application. The principle of operation of the Mark3 Moisture Analyzers is loss on drying. The instrument finalizes on a programmed rate of weight change compared to the actual initial weight of the sample over a programmed window of time. They feature a built-in temperature and weight calibration function to ensure advanced moisture analysis. The modular Mark3 with its add-ons of up to four heating units provides an unequaled flexibility and enables even parallel measurements of different samples.
  • Economical and reliable alternative to the Karl-Fischer titration and drying oven methods
  • Intuitive operation – With a learning curve of a few minutes
  • The user starts the measurement by pressing just one button
  • Robust and fast, optimal for routine operation
  • Multi-module operation enables parallel measurements of up to 4 different samples on one single device
  • An extensive range of security settings guarantees reliable results and 100% traceability
Mark3 HP Moisture Analyzer
A flexible moisture analyzer system for labs and production facilities. Its optional modular add-ons provide the best solution for every measuring task. Its high-resolution weighing system also makes the Mark3 HP suitable for precisely analyzing the moisture and water content of very dry samples. This feature makes it a genuine alternative to Karl-Fisher titration.