Norway
Order Entry
Baicalin
undefined
Baicalin
  • Appearance:
    White to yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    194 to 205 °C
  • HPLC:
    ≥94 %
  • Specific optical rotation:
    -80° to -90° (20 °C, 589 nm) (1% in DMSO)
  •  
    Pack type
    Pk
  • ACRO464900010
    464900010
    Glass bottle
    1 g
    1 * 1 g
    1 * 1 g - kr 505,00
    kr 505,00
    1 * 1 g
       kr 505,00
  • ACRO464900050
    464900050
    Glass bottle
    5 g
    1 * 5 g
    1 * 5 g - kr 1 100,00
    kr 1 100,00
    1 * 5 g
       kr 1 100,00