Quick response kit, chemical, HazWik®
Leverandør: Brady
116-0940EA 680 NOK
116-0940
Quick response kit, chemical, HazWik®
Materialer for håndtering av søl
Get fast and effective spill clean-up with a HAZWIK® quick response kit.
  • Compact, single-use kit that is ideal for use on small spills in the lab
  • Quickly cleans up hazardous chemical spills
  • Visual instructions on the package
It contains everything needed for emergency spills in a convenient 'grab and go' style bag.
Leveringsinformasjon: Contains PPE, tools, absorbent pads, acid and base neutralisers, wipes and disposable materials.