Norway
Order Entry
Tris(dimethylamino)phosphine
Flammable
Health
Tris(dimethylamino)phosphine
Catalog # APOSOR0499-25G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Health
Tris(dimethylamino)phosphine
Catalog # APOSOR0499-25G
Supplier:  Apollo Scientific
CAS Number:  
Price
kr 2 750,00
Log in for availability
Unit of Measure 1 * 25 g

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :163.2 g/mol
Boiling Pt:162…164 °C
Melting Pt:–44 °C
Density:0.898 g/cm³ (25 °C)
Flash Pt:37 °C
UN:1993
ADR:3,III
 :GHS02
 :GHS08
Storage Temperature:Ambient
CAS Number:1608-26-0
Formula:C₆H₁₈N₃P
MDL Number:MFCD00008301