3-Bromo-9-ethylcarbazole

Warning

APOSOR303353-5GEA 0 NOK
APOSOR303353-5G APOSOR303353-1G
3-Bromo-9-ethylcarbazole
3-Bromo-9-ethylcarbazole
Formel: C₁₄H₁₂BrN
MW: 274,1558 g/mol
Smeltepunkt: 84 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00092392
CAS-nummer: 57102-97-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR