o-Anisidinium chloride 98%
Leverandør: Apollo Scientific
APOSOR904667-100GEA
0
NOK
APOSOR904667-100G
APOSOR904667-25G
o-Anisidinium chloride 98%
o-Anisidinium chloride
Formel:
C₇H₁₀ClNO MW: 159,6134 g/mol Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00054334 CAS-nummer: 134-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...