Norway
Order Entry
J.T.Baker® BAKER INSTRA-ANALYZED®, Niobium Plasma Standards
J.T.Baker® BAKER INSTRA-ANALYZED®, Niobium Plasma Standards
 :  Avantor
J.T.Baker® BAKER INSTRA-ANALYZED®, Niobium Plasma Standards
 :  Avantor
  •  
    Element
    Symbol
    Concentration
    Matrix
    Pk
  • 5771.0100
    Niobium
    Nb
    1000 mg/L
    H₂O/tr HF
    100 mL
    1 * 100 mL
    1 * 100 mL
  • 5760.0100
    Niobium
    Nb
    10000 mg/L
    H₂O/tr HF
    100 mL
    1 * 100 mL
    1 * 100 mL