About this item
Ultra-clear polypropylene, graduated, autoclavable, and freezable microcentrifuge tubes with flat caps. Amber coloured styles are available for use with light-sensitive substances.
- Every lot is tested for protease, Human DNA, ATP, and RNase/DNase
- Endotoxin-free (non-pyrogenic)
- Supplied with flat, easy-open, attached snap caps
- Side graduations and frosted writing surface
- Polypropylene USP Class VI resin
- Withstands ultra-high centrifuging up to 40000×g
- Freezable to –80 °C
The ability to extract DNA from samples and then amplify that DNA with reliable results is an important part of lab work. SuperSpin® tubes are designed with this in mind. They can be trusted to spin at high rates, and can be opened and closed easily many times to add or remove reagents. The interior surfaces are ultra-smooth with low retention. They are clear enough to be used in a spectrophotometer and have no extractables at the wavelength ranges used to detect DNA.
Lab testing confirms the superior performance of SuperSpin® tubes in light transmission compared to popular brands. They are more than twice as effective at transmitting light than most brands of microcentrifuge tubes.
Product Information
- Catalog #Supplier #CapacityColourTypeRCF max.PackedAvailabilityPrice
- SuperSpin® microcentrifuge tubesSuperSpin® microcentrifuge tubes3016-871-000-93016-871-000-91,5 mlAmberAttached snapcap, non sterile40000×gResealable bagsLog in for availability1 * 500 Itemskr 680,00Supplier Article Number: 3016-871-000-9Local Article Number: LBCN3016-871-000-9Specifications:
Specifications
Environmentally Preferable - SuperSpin® microcentrifuge tubesSuperSpin® microcentrifuge tubes3016-879-000-93016-879-000-91,5 mlAssorted coloursAttached snapcap, non sterile40000×gResealable bagsLog in for availability1 * 500 Itemskr 795,00Supplier Article Number: 3016-879-000-9Local Article Number: LBCN3016-879-000-9Specifications:
Specifications
Environmentally Preferable - SuperSpin® microcentrifuge tubesSuperSpin® microcentrifuge tubes3016-877-000-93016-877-000-91,5 mlBlueAttached snapcap, non sterile40000×gResealable bagsLog in for availability1 * 500 Itemskr 680,00Supplier Article Number: 3016-877-000-9Local Article Number: LBCN3016-877-000-9Specifications:
Specifications
Environmentally Preferable - SuperSpin® microcentrifuge tubesSuperSpin® microcentrifuge tubes3016-870-000-93016-870-000-91,5 mlClearAttached snapcap, non sterile40000×gResealable bagsLog in for availability1 * 500 Itemskr 660,00Supplier Article Number: 3016-870-000-9Local Article Number: LBCN3016-870-000-9Specifications:
Specifications
Environmentally Preferable - SuperSpin® microcentrifuge tubesSuperSpin® microcentrifuge tubes3016-878-000-93016-878-000-91,5 mlGreenAttached snapcap, non sterile40000×gResealable bagsLog in for availability1 * 500 Itemskr 680,00Supplier Article Number: 3016-878-000-9Local Article Number: LBCN3016-878-000-9Specifications:
Specifications
Environmentally Preferable - SuperSpin® microcentrifuge tubesSuperSpin® microcentrifuge tubes3016-873-000-93016-873-000-91,5 mlOrangeAttached snapcap, non sterile40000×gResealable bagsLog in for availability1 * 500 Itemskr 680,00Supplier Article Number: 3016-873-000-9Local Article Number: LBCN3016-873-000-9Specifications:
Specifications
Environmentally Preferable - SuperSpin® microcentrifuge tubesSuperSpin® microcentrifuge tubes3016-876-000-93016-876-000-91,5 mlPurpleAttached snapcap, non sterile40000×gResealable bagsLog in for availability1 * 500 Itemskr 685,00Supplier Article Number: 3016-876-000-9Local Article Number: LBCN3016-876-000-9Specifications:
Specifications
Environmentally Preferable - SuperSpin® microcentrifuge tubesSuperSpin® microcentrifuge tubes3016-874-000-93016-874-000-91,5 mlRedAttached snapcap, non sterile40000×gResealable bagsLog in for availability1 * 500 Itemskr 680,00Supplier Article Number: 3016-874-000-9Local Article Number: LBCN3016-874-000-9Specifications:
Specifications
Environmentally Preferable - SuperSpin® microcentrifuge tubesSuperSpin® microcentrifuge tubes3016-872-000-93016-872-000-91,5 mlYellowAttached snapcap, non sterile40000×gResealable bagsLog in for availability1 * 500 Itemskr 685,00Supplier Article Number: 3016-872-000-9Local Article Number: LBCN3016-872-000-9Specifications:
Specifications
Environmentally Preferable