Norway
Order Entry
Chu N6 basal salt mixture
Chu N6 basal salt mixture
  •  
    Pk
  • Chu N6 basal salt mixture 10×1 L
    Chu N6 basal salt mixture 10×1 L
    APOSPMC416A-10X1L
    PMC416A-10X1L
    10
    1 * 10 Items
    1 * 10 Items
  • Chu N6 basal salt mixture 10 L
    Chu N6 basal salt mixture 10 L
    APOSPMC416B-10L
    PMC416B-10L
    10 L
    1 * 10 L
    1 * 10 L - kr 270,00
    kr 270,00
    1 * 10 L
       kr 270,00
  • Chu N6 basal salt mixture 50 L
    Chu N6 basal salt mixture 50 L
    APOSPMC416C-50L
    PMC416C-50L
    50 L
    1 * 50 L
    1 * 50 L