Incubators, IF/IFplus (forced convection)
Supplier: MEMMERT
Certificates
About this item
Specifications
- Catalog No:
- MEMMIF110M
- 390-0642
- MEMMIF75M
- MEMMIF75MPLUS
- 390-0633
- 390-0629
- 390-0628
- 390-0637
- MEMMIF55MPLUS
- MEMMIF450MPLUS
- MEMMIF55M
- 390-0632
- MEMMIF260M
- 390-0640
- MEMMIF160M
- MEMMIF260MPLUS
- MEMMIF30MPLUS
- 390-0643
- 390-0635
- 390-0639
- 390-0641
- 390-0631
- MEMMIF750MPLUS
- MEMMIF110MPLUS
- 390-0636
- MEMMIF450M
- MEMMIF160MPLUS
- 390-0638
- 390-0630
- 390-0634
- MEMMIF750M
- MEMMIF30M
- Model:
- IF110
- IF750
- IF75
- IF75
- IF260
- IF110
- IF110
- IF450
- IF55
- IF450
- IF55
- IF260
- IF260
- IF75
- IF160
- IF260
- IF30
- IF75
- IF30
- IF55
- IF750
- IF160
- IF750
- IF110
- IF450
- IF450
- IF160
- IF55
- IF160
- IF30
- IF750
- IF30
- Temperature fluctuation (time):
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- ≤±0,2 °C @ 37 °C
- Temperature variation (spatial):
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- ≤± 0,3 °C @ 37 °C
- Capacity:
- 108 L
- 749 L
- 74 L
- 74 L
- 256 L
- 108 L
- 108 L
- 449 L
- 53 L
- 449 L
- 53 L
- 256 L
- 256 L
- 74 L
- 161 L
- 256 L
- 32 L
- 74 L
- 32 L
- 53 L
- 749 L
- 161 L
- 749 L
- 108 L
- 449 L
- 449 L
- 161 L
- 53 L
- 161 L
- 32 L
- 749 L
- 32 L
- Convection type:
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Max. load per shelf:
- 20 kg
- 30 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 30 kg
- 20 kg
- 30 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 30 kg
- 20 kg
- 30 kg
- 20 kg
- 30 kg
- 30 kg
- 20 kg
- 20 kg
- 20 kg
- 20 kg
- 30 kg
- 20 kg
- Power consumption:
- 1400 W
- 2000 W
- 1250 W
- 1250 W
- 1700 W
- 1400 W
- 1400 W
- 1800 W
- 1000 W
- 1800 W
- 1000 W
- 1700 W
- 1700 W
- 1250 W
- 1600 W
- 1700 W
- 800 W
- 1250 W
- 800 W
- 1000 W
- 2000 W
- 1600 W
- 2000 W
- 1400 W
- 1800 W
- 1800 W
- 1600 W
- 1000 W
- 1600 W
- 800 W
- 2000 W
- 800 W
- Temperature range:
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- RT* +10…80 °C
- Weight:
- 72 kg
- 192 kg
- 64 kg
- 64 kg
- 96 kg
- 72 kg
- 72 kg
- 160 kg
- 55 kg
- 160 kg
- 55 kg
- 96 kg
- 96 kg
- 64 kg
- 80 kg
- 96 kg
- 44 kg
- 64 kg
- 44 kg
- 55 kg
- 192 kg
- 80 kg
- 192 kg
- 72 kg
- 160 kg
- 160 kg
- 80 kg
- 55 kg
- 80 kg
- 44 kg
- 192 kg
- 44 kg
Specifications
IF and IFplus models with forced convection
Product Information
- Catalog #Supplier #TypeShelves supplied (max.)W×D×H int.W×D×H ext.PkAvailabilityPrice
- 390-0642IF750PLUSIF750plus with TwinDISPLAY0 (14)1040×600×1200 mm1224×784×1726 mm11 * 1 Items
Accessories
Specifications
ModelIF750Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity749 LConvection typeForced convectionMax. load per shelf30 kgPower consumption2000 WTemperature rangeRT* +10…80 °CWeight192 kg - 390-0634IF30IF30 with SingleDISPLAY0 (3)400×250×320 mm585×434×707 mm11 * 1 Itemskr 22 800,00
Accessories
Specifications
ModelIF30Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity32 LConvection typeForced convectionMax. load per shelf20 kgPower consumption800 WTemperature rangeRT* +10…80 °CWeight44 kg - 390-0635IF30PLUSIF30plus with TwinDISPLAY0 (3)400×250×320 mm585×434×707 mm11 * 1 Itemskr 33 300,00
Accessories
Specifications
ModelIF30Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity32 LConvection typeForced convectionMax. load per shelf20 kgPower consumption800 WTemperature rangeRT* +10…80 °CWeight44 kg - 390-0638IF55IF55 with SingleDISPLAY0 (4)400×330×400 mm585×514×787 mm11 * 1 Itemskr 29 550,00
Accessories
Specifications
ModelIF55Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity53 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1000 WTemperature rangeRT* +10…80 °CWeight55 kg - 390-0639IF55PLUSIF55plus with TwinDISPLAY0 (4)400×330×400 mm585×514×787 mm11 * 1 Itemskr 39 800,00
Accessories
Specifications
ModelIF55Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity53 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1000 WTemperature rangeRT* +10…80 °CWeight55 kg - 390-0640IF75IF75 with SingleDISPLAY0 (6)400×330×560 mm585×514×947 mm11 * 1 Itemskr 32 675,00
Accessories
Specifications
ModelIF75Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity74 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1250 WTemperature rangeRT* +10…80 °CWeight64 kg - 390-0643IF75PLUSIF75plus with TwinDISPLAY0 (6)400×330×560 mm585×514×947 mm11 * 1 Itemskr 43 875,00
Accessories
Specifications
ModelIF75Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity74 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1250 WTemperature rangeRT* +10…80 °CWeight64 kg - 390-0628IF110IF110 with SingleDISPLAY0 (5)560×400×480 mm745×584×867 mm11 * 1 Itemskr 38 325,00
Accessories
Specifications
ModelIF110Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity108 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1400 WTemperature rangeRT* +10…80 °CWeight72 kg - 390-0629IF110PLUSIF110plus with TwinDISPLAY0 (5)560×400×480 mm745×584×867 mm11 * 1 Itemskr 48 600,00
Accessories
Specifications
ModelIF110Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity108 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1400 WTemperature rangeRT* +10…80 °CWeight72 kg - 390-0630IF160IF160 with SingleDISPLAY0 (8)560×400×720 mm745×584×1107 mm11 * 1 Itemskr 48 350,00
Accessories
Specifications
ModelIF160Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity161 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1600 WTemperature rangeRT* +10…80 °CWeight80 kg - 390-0631IF160PLUSIF160plus with TwinDISPLAY0 (8)560×400×720 mm745×584×1107 mm11 * 1 Itemskr 61 325,00
Accessories
Specifications
ModelIF160Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity161 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1600 WTemperature rangeRT* +10…80 °CWeight80 kg - 390-0633IF260PLUSIF260plus with TwinDISPLAY0 (9)640×500×800 mm824×684×1186 mm11 * 1 Itemskr 73 925,00
Accessories
Specifications
ModelIF260Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity256 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1700 WTemperature rangeRT* +10…80 °CWeight96 kg - 390-0636IF450IF450 with SingleDISPLAY0 (8)1040×600×720 mm1224×784×1247 mm11 * 1 Itemskr 79 400,00
Accessories
Specifications
ModelIF450Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity449 LConvection typeForced convectionMax. load per shelf30 kgPower consumption1800 WTemperature rangeRT* +10…80 °CWeight160 kg - 390-0637IF450PLUSIF450plus with TwinDISPLAY0 (8)1040×600×720 mm1224×784×1247 mm11 * 1 Items
Accessories
Specifications
ModelIF450Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity449 LConvection typeForced convectionMax. load per shelf30 kgPower consumption1800 WTemperature rangeRT* +10…80 °CWeight160 kg - 390-0632IF260IF260 with SingleDISPLAY0 (9)640×500×800 mm824×684×1186 mm11 * 1 Itemskr 62 825,00
Accessories
Specifications
ModelIF260Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity256 LConvection typeForced convectionMax. load per shelf20 kgPower consumption1700 WTemperature rangeRT* +10…80 °CWeight96 kg - 390-0641IF750IF750 with SingleDISPLAY0 (14)1040×600×1200 mm1224×784×1726 mm11 * 1 Items
Accessories
Specifications
ModelIF750Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity749 LConvection typeForced convectionMax. load per shelf30 kgPower consumption2000 WTemperature rangeRT* +10…80 °CWeight192 kg
IFm and IFmplus models with forced convection, Class 1 medical device
Product Information
- Catalog #Supplier #TypeShelves supplied (max.)W×D×H int.W×D×H ext.PkAvailabilityPrice
- MEMMIF110MIF110MIF110m with SingleDISPLAY0 (5)560×400×480 mm745×584×867 mm11 * 1 Itemskr 42 500,00
Accessories
Specifications
ModelIF110Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity108 LConvection typeMax. load per shelf20 kgPower consumption1400 WTemperature rangeRT* +10…80 °CWeight72 kg - MEMMIF75MIF75MIF75m with SingleDISPLAY0 (6)400×330×560 mm585×514×947 mm11 * 1 Itemskr 36 200,00
Accessories
Specifications
ModelIF75Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity74 LConvection typeMax. load per shelf20 kgPower consumption1250 WTemperature rangeRT* +10…80 °CWeight64 kg - MEMMIF75MPLUSIF75MPLUSIF75mplus with TwinDISPLAY0 (6)400×330×560 mm585×514×947 mm11 * 1 Itemskr 48 475,00
Accessories
Specifications
ModelIF75Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity74 LConvection typeMax. load per shelf20 kgPower consumption1250 WTemperature rangeRT* +10…80 °CWeight64 kg - MEMMIF30MPLUSIF30MPLUSIF30mplus with TwinDISPLAY0 (3)400×250×320 mm585×434×707 mm11 * 1 Itemskr 36 700,00
Accessories
Specifications
ModelIF30Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity32 LConvection typeMax. load per shelf20 kgPower consumption800 WTemperature rangeRT* +10…80 °CWeight44 kg - MEMMIF55MIF55MIF55m with SingleDISPLAY0 (4)400×330×400 mm585×514×787 mm11 * 1 Itemskr 32 575,00
Accessories
Specifications
ModelIF55Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity53 LConvection typeMax. load per shelf20 kgPower consumption1000 WTemperature rangeRT* +10…80 °CWeight55 kg - MEMMIF55MPLUSIF55MPLUSIF55mplus with TwinDISPLAY0 (4)400×330×400 mm585×514×787 mm11 * 1 Itemskr 43 850,00
Accessories
Specifications
ModelIF55Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity53 LConvection typeMax. load per shelf20 kgPower consumption1000 WTemperature rangeRT* +10…80 °CWeight55 kg - MEMMIF110MPLUSIF110MPLUSIF110mplus with TwinDISPLAY0 (5)560×400×480 mm745×584×867 mm11 * 1 Itemskr 53 775,00
Accessories
Specifications
ModelIF110Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity108 LConvection typeMax. load per shelf20 kgPower consumption1400 WTemperature rangeRT* +10…80 °CWeight72 kg - MEMMIF160MPLUSIF160MPLUSIF160mplus with TwinDISPLAY0 (8)560×400×720 mm745×584×1107 mm11 * 1 Itemskr 67 775,00
Accessories
Specifications
ModelIF160Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity161 LConvection typeMax. load per shelf20 kgPower consumption1600 WTemperature rangeRT* +10…80 °CWeight80 kg - MEMMIF450MIF450MIF450m with SingleDISPLAY0 (8)1040×600×720 mm1224×784×1247 mm11 * 1 Items
Accessories
Specifications
ModelIF450Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity449 LConvection typeMax. load per shelf30 kgPower consumption1800 WTemperature rangeRT* +10…80 °CWeight160 kg - MEMMIF450MPLUSIF450MPLUSIF450mplus with TwinDISPLAY0 (8)1040×600×720 mm1224×784×1247 mm11 * 1 Items
Accessories
Specifications
ModelIF450Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity449 LConvection typeMax. load per shelf30 kgPower consumption1800 WTemperature rangeRT* +10…80 °CWeight160 kg - MEMMIF260MIF260MIF260m with SingleDISPLAY0 (9)640×500×800 mm824×684×1186 mm11 * 1 Itemskr 69 475,00
Accessories
Specifications
ModelIF260Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity256 LConvection typeMax. load per shelf20 kgPower consumption1700 WTemperature rangeRT* +10…80 °CWeight96 kg - MEMMIF160MIF160MIF160m with SingleDISPLAY0 (8)560×400×720 mm745×584×1107 mm11 * 1 Itemskr 53 525,00
Accessories
Specifications
ModelIF160Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity161 LConvection typeMax. load per shelf20 kgPower consumption1600 WTemperature rangeRT* +10…80 °CWeight80 kg - MEMMIF260MPLUSIF260MPLUSIF260mplus with TwinDISPLAY0 (9)640×500×800 mm824×684×1186 mm11 * 1 Items
Accessories
Specifications
ModelIF260Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity256 LConvection typeMax. load per shelf20 kgPower consumption1700 WTemperature rangeRT* +10…80 °CWeight96 kg - MEMMIF750MPLUSIF750MPLUSIF750mplus with TwinDISPLAY0 (14)1040×600×1200 mm1224×784×1726 mm11 * 1 Items
Accessories
Specifications
ModelIF750Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity749 LConvection typeMax. load per shelf30 kgPower consumption2000 WTemperature rangeRT* +10…80 °CWeight192 kg - MEMMIF30MIF30MIF30m with SingleDISPLAY0 (3)400×250×320 mm585×434×707 mm11 * 1 Itemskr 25 150,00
Accessories
Specifications
ModelIF30Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity32 LConvection typeMax. load per shelf20 kgPower consumption800 WTemperature rangeRT* +10…80 °CWeight44 kg - MEMMIF750MIF750MIF750m with SingleDISPLAY0 (14)1040×600×1200 mm1224×784×1726 mm11 * 1 Items
Accessories
Specifications
ModelIF750Environmentally PreferableTemperature fluctuation (time)≤±0,2 °C @ 37 °CTemperature variation (spatial)≤± 0,3 °C @ 37 °CCapacity749 LConvection typeMax. load per shelf30 kgPower consumption2000 WTemperature rangeRT* +10…80 °CWeight192 kg
Stainless Steel Grids
Product Information
- Catalog #SeqDescriptionForPkPriceAvailabilityPrice
Perforated Stainless Steel Shelves
Product Information
- Catalog #SeqDescriptionForPkPriceAvailabilityPrice
Non-perforated Stainless Steel Trays with 15 mm Rim
Product Information
- Catalog #SeqDescriptionForPkPriceAvailabilityPrice
Bottom Drip Tray
Product Information
- Catalog #SeqDescriptionTypePkPriceAvailabilityPrice
Wall Bracket: Tubular Frame for Wall Mounting
Product Information
- Catalog #SeqTypeDescriptionPkPriceAvailabilityPrice
Port, 14 mm Ø
Product Information
- Catalog #SeqDescriptionForPkAvailabilityPrice
- 05Port, Ø 14 mm, can be closed by flap, custom positions in the back wallIN2601Log in for availability
Subframes
Product Information
- Catalog #SeqDescriptionPkAvailabilityPrice