Tissue RNA extraction kit, Precellys®
Leverandør: Bertin
Precellys®
BERTD05711.50EAEA 5150 NOK
BERTD05711.50EA
Tissue RNA extraction kit, Precellys®
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
Precellys® homogenisers and nucleic acid extraction kits offer an optimised solution to purify RNA from your tissue samples.
  • Premium quality: High yields of purified RNA
  • Optimised Precellys® workflow from tissue to nucleic acid
  • Easy to use: Spin column technology
  • Versatile: Single kits for multiple tissue types
Our Precellys® tissue RNA extraction kit allow for the extraction and purification of DNA from a variety of tissue samples such as heart, liver, ear or tail. The silica based mini column technology delivers highly pure nucleic acids that are free from protein and other contaminants and compatible with a large range of downstream applications such as PCR, RT-qPCR, sequencing, genotyping, Southern blot and Northern analysis.
Leveringsinformasjon: The kits include a comprehensive user guide with optimised protocols for tissue homogenisation and nucleic acid purification.
NB: Store at room temperature. For research use only.