Norway
Order Entry
VWR®, Vial Racks
undefined
VWR®, Vial Racks
  •  
    Hole Ø
    No. of Holes
    W×D×H
    Pk
  • Vial rack for 4 ml vials, transparent, acryl
    Vial rack for 4 ml vials, transparent, acryl
    548-0192A
    15,2 mm
    40
    175,8×115,5×20 mm
    1
    1 * 1 Items
    1 * 1 Items - kr 1 350,00
    kr 1 350,00
    1 * 1 Items
       kr 1 350,00
    Specifications
    Environmentally Preferable
  • Vial rack for 4 ml vials, blue, PP, stackable
    Vial rack for 4 ml vials, blue, PP, stackable
    548-1520A
    14,9 mm
    50
    238×120×28 mm
    1
    1 * 1 Items
    1 * 1 Items - kr 425,00
    kr 425,00
    1 * 1 Items
       kr 425,00
    Specifications
    Environmentally Preferable
  • Vial rack for 1,5 and 2 ml vials, transparent, acryl
    Vial rack for 1,5 and 2 ml vials, transparent, acryl
    548-0131A
    12,2 mm
    50
    173×95×20 mm
    1
    1 * 1 Items
    1 * 1 Items - kr 2 100,00
    kr 2 100,00
    1 * 1 Items
       kr 2 100,00
    Specifications
    Environmentally Preferable