Headspace ND20 vials with crimp neck manufactured from 1st hydrolytic class borosilicate glass in both flat bottom and round bottom style. They can be purchased with a variety of aluminium seal styles and septa materials.
- High performance caps/septa
- Available with flat DIN crimp neck for tighter seal
- Thicker wall 1,2 mm withstand higher internal pressure
Headspace ND20 vials are available in three different sizes (5, 10 and 20 ml) in amber or clear version. For use in a broad variety of instrument platforms the vials are available with round or a flat bottom. During the heating process a rounded bottom vial is more resistant and robust to high pressure. Broad range of different crimp caps (center hole, tear off, pressure release or magnetic) mostly available with assembled stopper or septa are engineered to prevent sample contamination while providing consistent, secure seals.
Vials
- CapacityBaseØ×HMaterialColourCap included
- Vials, bevelledVials, bevelled548-0057A5,00 mlRounded bottom (HS bottom)22×38,2 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 6 100,00
Accessories
Specifications
Environmentally Preferable - VialsVials548-0611A5,00 mlFlat bottom20×38 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 8 575,00
Accessories
Specifications
Environmentally Preferable - Vials, bevelledVials, bevelled548-2365A5,00 mlFlat bottom21,7×38,2 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 5 325,00
Accessories
Specifications
Environmentally Preferable - Vials, DIN, long neckVials, DIN, long neck548-0090A10,00 mlFlat bottom22,5×46 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 4 675,00
Accessories
Specifications
Environmentally Preferable - Vials, DINVials, DIN548-0133A10,00 mlRounded bottom (HS bottom)22,5×46 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 4 525,00
Accessories
Specifications
Environmentally Preferable - VialsVials548-0612A10,00 mlFlat bottom20×54,4 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 6 900,00
Accessories
Specifications
Environmentally Preferable - Vials, DINVials, DIN548-0799A10,00 mlRounded bottom (HS bottom)22,5×46 mmBorosilicate glass 5.1AmberNo1 * 1.000 Itemskr 5 950,00
Accessories
Specifications
Environmentally Preferable - Vials, bevelledVials, bevelled548-1990A10,00 mlRounded bottom (HS bottom)22,5×46 mmBorosilicate glass 5.1ClearNo1 * 100 Itemskr 485,00
Accessories
Specifications
Environmentally Preferable - Vials, bevelledVials, bevelled548-0055A20,00 mlRounded bottom (HS bottom)23×75,5 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 5 050,00
Accessories
Specifications
Environmentally Preferable - Vials, DIN, long neckVials, DIN, long neck20,00 mlFlat bottom22,5×75,5 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 6 775,00
Accessories
Specifications
Environmentally Preferable - Vials, DIN, long neckVials, DIN, long neck548-0151A20,00 mlRounded bottom (HS bottom)22,5×75,5 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 10 575,00
Accessories
Specifications
Environmentally Preferable - Vials with write-on patch, bevelledVials with write-on patch, bevelled548-0610A20,00 mlRounded bottom (HS bottom)23×75,5 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 7 350,00
Accessories
Specifications
Environmentally Preferable - Vials, special crimp neckVials, special crimp neck548-0613A20,00 mlRounded bottom (HS bottom)22,75×75,5 mm1 * 100 Itemskr 1 425,00
Accessories
Specifications
Environmentally Preferable - Vials, bevelledVials, bevelled548-0614A20,00 mlRounded bottom (HS bottom)23×75,5 mmBorosilicate glass 5.1AmberNo1 * 1.000 Itemskr 7 000,00
Accessories
Specifications
Environmentally Preferable - Vials, DIN, long neckVials, DIN, long neck548-0798A20,00 mlRounded bottom (HS bottom)22,5×75,5 mmBorosilicate glass 5.1AmberNo1 * 1.000 Itemskr 8 125,00
Accessories
Specifications
Environmentally Preferable - Vials with write-on patch, DIN, long neckVials with write-on patch, DIN, long neck548-0891A20,00 mlRounded bottom (HS bottom)22,5×75,5 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 8 125,00
Accessories
Specifications
Environmentally Preferable - Vials, DINVials, DIN548-2092A50,00 mlFlat bottom29×101 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 10 850,00
Accessories
Specifications
Environmentally Preferable - Vials, bevelledVials, bevelled20,00 mlRounded bottom (HS bottom)22,5×75,5 mmBorosilicate glass 5.1ClearNo1 * 100 Itemskr 550,00
Accessories
Specifications
Environmentally Preferable - Vials, bevelledVials, bevelled548-1994A20,00 mlFlat bottom22,75×75,5 mmBorosilicate glass 5.1ClearNo1 * 1.000 Itemskr 5 475,00
Accessories
Specifications
Environmentally Preferable
Vial kits, unassembled (2 in 1)
- CapacityBaseØ×HMaterialColourCap included
- Vial kits, bevelledVial kits, bevelled548-2085A20,00 mlRounded bottom (HS bottom)23×75,5 mmBorosilicate glass 5.1ClearYes1 * 1.000 Itemskr 11 975,00
Accessories
Specifications
Environmentally Preferable - Vial kits, bevelledVial kits, bevelled5,00 mlFlat bottom21,7×38,2 mmBorosilicate glass 5.1ClearYes1 * 1.000 Itemskr 15 250,00
Accessories
Specifications
Environmentally Preferable
Crimp caps, open top
- SeqDescriptionHardnessSeptumThicknessCap colourCap SizeMaterial
- 03Crimp caps with septa, open top50° shore AGrey bromo-butyl/PTFE, completely laminated3,0 mmSilverND20Aluminium
Crimp caps, open top, pressure release
- SeqDescriptionHardnessSeptumThicknessCap colourCap SizeMaterial
- 21Crimp caps with septa, open top, pressure release, high temperature quality, high purity45° shore AWhite silicone/beige PTFE3,2 mmSilverND20Aluminium
Crimp caps, open top, magnetic
- SeqDescriptionHardnessSeptumThicknessCap colourCap SizeMaterial
- 29Crimp caps with septa, open top, magnetic, high purity, 5 mm hole45° shore ATransparent blue silicone/white PTFE, completely laminated3,0 mmGoldND20Steel
Crimp caps, centre tear off
- SeqDescriptionHardnessSeptumThicknessCap colourCap SizeMaterial
- 46Crimp caps with septa, centre tear off50° shore APharma-fix bromo-butyl/PTFE3,0 mmSilverND20Aluminium
Crimp caps, complete tear off
- SeqDescriptionHardnessSeptumThicknessCap colourCap SizeMaterial
- 55Crimp caps with septa, complete tear off50° shore APharma-fix bromo-butyl/PTFE3,0 mmSilverND20Aluminium
Snap caps, push on
- SeqDescriptionHardnessSeptumThicknessCap colourCap SizeMaterial
- 65Snap caps, push on, for DIN neck55° shore ABeige butyl/grey PTFE1,3 mmTransparentND20PE
Stoppers
- SeqDescriptionHardnessSeptumThicknessCap SizePk
- 01Stopper50° shore ADark grey bromo-butyl3,0 mmND201.000
Septa ND20
- SeqSeptumHardnessThicknessPk
- 01Red butyl/grey PTFE50° shore A1,3 mm1.000