Masterflex® Metal Metering Valves, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Choose from different metal materials based on your application.
- Brass and 316 Stainless Steel material options available
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX03214-84
- MFLX03218-84
- MFLX03214-83
- MFLX03218-85
- MFLX03214-85
- MFLX03214-86
- MFLX03214-91
- MFLX03214-92
- MFLX03214-93
- MFLX03214-94
- MFLX03214-95
- MFLX03214-96
- MFLX03214-81
- MFLX03218-76
- MFLX03214-82
- MFLX03218-77
- MFLX03218-79
- MFLX03218-81
- MFLX03218-83
- MFLX03218-87
- MFLX03218-78
- Model:
- 03214-84
- 03218-84
- 03214-83
- 03218-85
- 03214-85
- 03214-86
- 03214-91
- 03214-92
- 03214-93
- 03214-94
- 03214-95
- 03214-96
- 03214-81
- 03218-76
- 03214-82
- 03218-77
- 03218-79
- 03218-81
- 03218-83
- 03218-87
- 03218-78
- Max. temperature:
- 81 °C (180 °F)
- 81 °C (180 °F)
- 81 °C (180 °F)
- 121 °C (250 °F)
- 81 °C (180 °F)
- 81 °C (180 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 81 °C (180 °F)
- 81 °C (180 °F)
- 81 °C (180 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 81 °C (180 °F)
- Orifice:
- 2,3622 mm (0,093")
- 3,048 mm (0,12")
- 1,0668 mm (0,042")
- 3,048 mm (0,12")
- 2,3622 mm (0,093")
- 2,3622 mm (0,093")
- 1,0668 mm (0,042")
- 1,0668 mm (0,042")
- 1,0668 mm (0,042")
- 2,3622 mm (0,093")
- 2,3622 mm (0,093")
- 2,3622 mm (0,093")
- 1,0668 mm (0,042")
- 1,3208 mm (0,052")
- 1,0668 mm (0,042")
- 1,3208 mm (0,052")
- 1,3208 mm (0,052")
- 2,0828 mm (0,082")
- 2,0828 mm (0,082")
- 3,048 mm (0,12")
- 1,3208 mm (0,052")
- Wetted materials:
- Brass
- Brass
- Brass
- Stainless Steel
- Brass
- Brass
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Brass
- Brass
- Brass
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Stainless Steel
- Brass
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Specifications
Product Information
- Catalog #Supplier #Connection TypeInlet portOutlet portPort SizeFlow patternAvailabilityPrice
- High-Resolution Brass Metering Valve, straight flow, 6 ml/minHigh-Resolution Brass Metering Valve, straight flow, 6 ml/min03214-81NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 1 300,00Supplier Article Number: 03214-81Local Article Number: MFLB03214-81Specifications:
More
Specifications
Model03214-81Environmentally PreferableMax. temperature81 °C (180 °F)Orifice1,0668 mm (0,042")Wetted materialsBrassWarranty1 year - High-Resolution Brass Metering Valve, straight flow, 12 ml/minHigh-Resolution Brass Metering Valve, straight flow, 12 ml/min03214-82NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 1 300,00Supplier Article Number: 03214-82Local Article Number: MFLB03214-82Specifications:
More
Specifications
Model03214-82Environmentally PreferableMax. temperature81 °C (180 °F)Orifice1,0668 mm (0,042")Wetted materialsBrassWarranty1 year - High-Resolution Brass Metering Valve, straight flow, 30 ml/minHigh-Resolution Brass Metering Valve, straight flow, 30 ml/min03214-83NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 1 300,00Supplier Article Number: 03214-83Local Article Number: MFLB03214-83Specifications:
More
Specifications
Model03214-83Environmentally PreferableMax. temperature81 °C (180 °F)Orifice1,0668 mm (0,042")Wetted materialsBrassWarranty1 year - High-Resolution Brass Metering Valve, straight flow, 70 ml/minHigh-Resolution Brass Metering Valve, straight flow, 70 ml/min03214-84NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 1 300,00Supplier Article Number: 03214-84Local Article Number: MFLB03214-84Specifications:
More
Specifications
Model03214-84Environmentally PreferableMax. temperature81 °C (180 °F)Orifice2,3622 mm (0,093")Wetted materialsBrassWarranty1 year - High-Resolution Brass Metering Valve, straight flow, 200 ml/minHigh-Resolution Brass Metering Valve, straight flow, 200 ml/min03214-85NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 1 300,00Supplier Article Number: 03214-85Local Article Number: MFLB03214-85Specifications:
More
Specifications
Model03214-85Environmentally PreferableMax. temperature81 °C (180 °F)Orifice2,3622 mm (0,093")Wetted materialsBrassWarranty1 year - High-Resolution Brass Metering Valve, straight flow, 650 ml/minHigh-Resolution Brass Metering Valve, straight flow, 650 ml/min03214-86NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 1 350,00Supplier Article Number: 03214-86Local Article Number: MFLB03214-86Specifications:
More
Specifications
Model03214-86Environmentally PreferableMax. temperature81 °C (180 °F)Orifice2,3622 mm (0,093")Wetted materialsBrassWarranty1 year - High-Resolution 316 SS Metering Valve, straight flow, 6 ml/minHigh-Resolution 316 SS Metering Valve, straight flow, 6 ml/min03214-91NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 2 200,00Supplier Article Number: 03214-91Local Article Number: MFLB03214-91Specifications:
More
Specifications
Model03214-91Environmentally PreferableMax. temperature121 °C (250 °F)Orifice1,0668 mm (0,042")Wetted materialsStainless SteelWarranty1 year - High-Resolution 316 SS Metering Valve, straight flow, 12 ml/minHigh-Resolution 316 SS Metering Valve, straight flow, 12 ml/min03214-92NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 2 200,00Supplier Article Number: 03214-92Local Article Number: MFLB03214-92Specifications:
More
Specifications
Model03214-92Environmentally PreferableMax. temperature121 °C (250 °F)Orifice1,0668 mm (0,042")Wetted materialsStainless SteelWarranty1 year - High-Resolution 316 SS Metering Valve, straight flow, 30 ml/minHigh-Resolution 316 SS Metering Valve, straight flow, 30 ml/min03214-93NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 2 200,00Supplier Article Number: 03214-93Local Article Number: MFLB03214-93Specifications:
More
Specifications
Model03214-93Environmentally PreferableMax. temperature121 °C (250 °F)Orifice1,0668 mm (0,042")Wetted materialsStainless SteelWarranty1 year - High-Resolution 316 SS Metering Valve, straight flow, 70 ml/minHigh-Resolution 316 SS Metering Valve, straight flow, 70 ml/min03214-94NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 2 200,00Supplier Article Number: 03214-94Local Article Number: MFLB03214-94Specifications:
More
Specifications
Model03214-94Environmentally PreferableMax. temperature121 °C (250 °F)Orifice2,3622 mm (0,093")Wetted materialsStainless SteelWarranty1 year - High-Resolution 316 SS Metering Valve, straight flow, 200 ml/minHigh-Resolution 316 SS Metering Valve, straight flow, 200 ml/min03214-95NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 2 200,00Supplier Article Number: 03214-95Local Article Number: MFLB03214-95Specifications:
More
Specifications
Model03214-95Environmentally PreferableMax. temperature121 °C (250 °F)Orifice2,3622 mm (0,093")Wetted materialsStainless SteelWarranty1 year - High-Resolution 316 SS Metering Valve, straight flow, 650 ml/minHigh-Resolution 316 SS Metering Valve, straight flow, 650 ml/min03214-96NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 2 200,00Supplier Article Number: 03214-96Local Article Number: MFLB03214-96Specifications:
More
Specifications
Model03214-96Environmentally PreferableMax. temperature121 °C (250 °F)Orifice2,3622 mm (0,093")Wetted materialsStainless SteelWarranty1 year - Brass Metering Valve, straight flow, 350 ml/minBrass Metering Valve, straight flow, 350 ml/min03218-76NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 775,00Supplier Article Number: 03218-76Local Article Number: MFLB03218-76Specifications:
More
Specifications
Model03218-76Environmentally PreferableMax. temperature81 °C (180 °F)Orifice1,3208 mm (0,052")Wetted materialsBrassWarranty1 year - 316 SS Metering Valve, straight flow, 350 ml/min316 SS Metering Valve, straight flow, 350 ml/min03218-77NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 1 300,00Supplier Article Number: 03218-77Local Article Number: MFLB03218-77Specifications:
More
Specifications
Model03218-77Environmentally PreferableMax. temperature121 °C (250 °F)Orifice1,3208 mm (0,052")Wetted materialsStainless SteelWarranty1 year - Brass Metering Valve, 90 deg flow, 350 ml/minBrass Metering Valve, 90 deg flow, 350 ml/min03218-78NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)90°Log in for availability1 * 1 Itemskr 800,00Supplier Article Number: 03218-78Local Article Number: MFLB03218-78Specifications:
More
Specifications
Model03218-78Environmentally PreferableMax. temperature81 °C (180 °F)Orifice1,3208 mm (0,052")Wetted materialsBrassWarranty1 year - 316 SS Metering Valve, 90 deg flow, 350 ml/min316 SS Metering Valve, 90 deg flow, 350 ml/min03218-79NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)90°Log in for availability1 * 1 Itemskr 1 325,00Supplier Article Number: 03218-79Local Article Number: MFLB03218-79Specifications:
More
Specifications
Model03218-79Environmentally PreferableMax. temperature121 °C (250 °F)Orifice1,3208 mm (0,052")Wetted materialsStainless SteelWarranty1 year - 316 SS Metering Valve, straight flow, 1200 ml/min316 SS Metering Valve, straight flow, 1200 ml/min03218-81NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 1 300,00Supplier Article Number: 03218-81Local Article Number: MFLB03218-81Specifications:
More
Specifications
Model03218-81Environmentally PreferableMax. temperature121 °C (250 °F)Orifice2,0828 mm (0,082")Wetted materialsStainless SteelWarranty1 year - 316 SS Metering Valve, 90 deg flow, 1200 ml/min316 SS Metering Valve, 90 deg flow, 1200 ml/min03218-83NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)90°Log in for availability1 * 1 Itemskr 1 325,00Supplier Article Number: 03218-83Local Article Number: MFLB03218-83Specifications:
More
Specifications
Model03218-83Environmentally PreferableMax. temperature121 °C (250 °F)Orifice2,0828 mm (0,082")Wetted materialsStainless SteelWarranty1 year - Brass Metering Valve, straight flow, 3500 ml/minBrass Metering Valve, straight flow, 3500 ml/min03218-84NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 775,00Supplier Article Number: 03218-84Local Article Number: MFLB03218-84Specifications:
More
Specifications
Model03218-84Environmentally PreferableMax. temperature81 °C (180 °F)Orifice3,048 mm (0,12")Wetted materialsBrassWarranty1 year - 316 SS Metering Valve, straight flow, 3500 ml/min316 SS Metering Valve, straight flow, 3500 ml/min03218-85NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)StraightLog in for availability1 * 1 Itemskr 1 300,00Supplier Article Number: 03218-85Local Article Number: MFLB03218-85Specifications:
More
Specifications
Model03218-85Environmentally PreferableMax. temperature121 °C (250 °F)Orifice3,048 mm (0,12")Wetted materialsStainless SteelWarranty1 year - 316 SS Metering Valve, 90 deg flow, 3500 ml/min316 SS Metering Valve, 90 deg flow, 3500 ml/min03218-87NPT(F)1/8" NPT(F)1/8" NPT(F)0,125 NPT(F)90°Log in for availability1 * 1 Itemskr 1 325,00Supplier Article Number: 03218-87Local Article Number: MFLB03218-87Specifications:
More
Specifications
Model03218-87Environmentally PreferableMax. temperature121 °C (250 °F)Orifice3,048 mm (0,12")Wetted materialsStainless SteelWarranty1 year