Poloxamer, Corning®
Supplier: Corning
Certificates
About this item
Poloxamer 188 is a surfactant used in cell culture to protect cells in suspension against possible damage during transfer, freezing and thawing, and stirring.
It is a non-ionic detergent with an average molecular weight of 8400Da consisting of a central polypropylene flanked by two polyethylene molecules.
Specifications
- Storage temperature:Refrigerate
Specifications
Related Information
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
- Poloxamer 188Supplier Article Number: 13-901-CILocal Article Number: MDTC13-901-CI
- Poloxamer 188Supplier Article Number: 61-161-RMLocal Article Number: MDTC61-161-RM