Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Norway
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
kr 0,00
Order Entry
Home
Standards
Organic Standards
1,6-anhydro-D-glucose, Reference Standard
1,6-anhydro-D-glucose, Reference Standard
1,6-anhydro-D-glucose, Reference Standard
1035300.
:
USP
:
1,6-anhydro-D-glucose, Reference Standard
Catalog # 1035300.
:
USP
:
: 1035300
: USPH1035300
kr 4 725,00
1 * 50 mg
-
+
Pack type:
Vial
Pk:
50 mg
Standards:
USP reference standard
CAS No.:
498-07-7
Description:
1,6-anhydro-D-glucose
Description:
1,6-anhydro-D-glucose