6-Methyl-4-phenylchromanone, Pharmaceutical Analytical Impurities
1A01060.
: USP
:
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:40546-94-9
- Description:6-Methyl-4-phenylchromanone
- Synonym(s):6-Methyl-4-phenylchroman-2-one
- Description:6-Methyl-4-phenylchromanone
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.